Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
dcun1d2
Entrez Gene ID
100145003
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 2
Gene Aliases
-
Chromosome Location
Chr.2: 112316073 - 112330644 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126565.1 NP_001120037.1 5 - 6 663 120
XM_012956436.2 XP_012811890.1 6 - 7 790 120
XM_012956437.2 XP_012811891.1 6 - 7 1383 120
XM_012956438.2 XP_012811892.1 5 - 6 1286 120
GenBank mRNA BC158344.1 - 5 - 6 663 120

View Full Product Details