Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
haus7
Entrez Gene ID
100145089
Gene Name
HAUS augmin like complex subunit 7
Gene Aliases
uchl5ip, uip1
Chromosome Location
Chr.7: 41956420 - 41978519 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126608.1 NP_001120080.1 3 - 4 332 111
GenBank mRNA BC158519.1 - 3 - 4 332 111

View Full Product Details