Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sntb2
Entrez Gene ID
100145613
Gene Name
syntrophin, beta 2 (dystrophin-associated protein A1, 59kDa, basic component 2)
Gene Aliases
-
Chromosome Location
Chr.4: 62145619 - 62159345 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127021.1 NP_001120493.1 3 - 4 952 100
GenBank mRNA BC161370.1 - 3 - 4 952 100

View Full Product Details