Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc35f2
Entrez Gene ID
100145019
Gene Name
solute carrier family 35 member F2
Gene Aliases
-
Chromosome Location
Chr.2: 156914888 - 156937415 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126571.1 NP_001120043.1 4 - 5 685 75
GenBank mRNA BC158377.1 - 4 - 5 685 75

View Full Product Details