Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zbtb8b
Entrez Gene ID
100145547
Gene Name
zinc finger and BTB domain containing 8B
Gene Aliases
znf916b
Chromosome Location
Chr.2: 55310675 - 55320566 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126981.1 NP_001120453.1 2 - 3 945 86
XM_012956491.2 XP_012811945.1 3 - 4 1147 86
XM_012956492.1 XP_012811946.1 2 - 3 1121 86
GenBank mRNA BC161234.1 - 2 - 3 945 86

View Full Product Details