Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dph1
Entrez Gene ID
100216186
Gene Name
diphthamide biosynthesis 1
Gene Aliases
dph1-ovca2, dph2l, dph2l1, ovca1
Chromosome Location
Chr.2: 27766906 - 27944321 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142155.1 NP_001135627.1 9 - 10 1221 101
XM_012956575.2 XP_012812029.1 9 - 10 1207 101
GenBank mRNA BC168479.1 - 9 - 10 1221 101
CT025515.2 - 9 - 10 1068 101

View Full Product Details