Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eln2
Entrez Gene ID
100217330
Gene Name
tropoelastin 2
Gene Aliases
-
Chromosome Location
Chr.9: 7003838 - 7026531 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142826.2 NP_001136298.1 23 - 24 1001 70
XM_012970194.2 XP_012825648.1 22 - 23 979 70
XM_012970195.2 XP_012825649.1 23 - 24 1021 70
XM_012970196.2 XP_012825650.1 23 - 24 1021 70
GenBank mRNA CU075851.1 - 23 - 24 1154 70
EU220008.1 - 23 - 24 1003 70

View Full Product Details