Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cldn5
Entrez Gene ID
448343
Gene Name
claudin 5 (transmembrane protein deleted in velocardiofacial syndrome)
Gene Aliases
awal, bec1, claudin-5, cpetrl1, tmvcf
Chromosome Location
Chr.1: 145813932 - 145816493 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006706.1 NP_001006707.1 1 - 1 546 94
GenBank mRNA BC075424.1 - 1 - 1 546 94

View Full Product Details