Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC100281180
Entrez Gene ID
100281180
Gene Name
gamma-glutamyltranspeptidase 1
Gene Aliases
ZEAMMB73_073980
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001154099.1 NP_001147571.1 - 1125 65
GenBank mRNA EU955837.1 - - 1125 65

View Full Product Details