Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
crs1
Entrez Gene ID
541870
Gene Name
chloroplast RNA splicing1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111538.1 NP_001105008.1 - 1771 82
GenBank mRNA AF290414.1 - - 1771 82
AF290415.1 - - 1262 82
AY109620.1 - - 1771 82

View Full Product Details