Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
sdh1
Entrez Gene ID
100283066
Gene Name
sorbitol dehydrogenase homolog1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001155968.1 NP_001149440.1 - 217 56
GenBank mRNA EU963336.1 - - 217 56

View Full Product Details