Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
sut2
Entrez Gene ID
100280251
Gene Name
sucrose transporter2
Gene Aliases
ZEAMMB73_722694
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001153179.1 NP_001146651.1 - 600 72
GenBank mRNA BT055860.1 - - 600 72

View Full Product Details