Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
phot1
Entrez Gene ID
541663
Gene Name
blue-light receptor phototropin 1
Gene Aliases
nph1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111416.1 NP_001104886.1 - 1661 71
GenBank mRNA AF033263.1 - - 1661 71

View Full Product Details