Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
lhcb6
Entrez Gene ID
542321
Gene Name
light harvesting chlorophyll a/b binding protein6
Gene Aliases
Lhcb6-1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111905.1 NP_001105375.1 - 848 103
GenBank mRNA U23190.1 - - 848 103

View Full Product Details