Gene Transcripts
- Gene Symbol
- Mir20a
- Entrez Gene ID
- 387139
- Gene Name
- microRNA 20a
- Gene Aliases
- Mir-20, Mirn20, Mirn20a, mir-20a, mmu-mir-20a
- Chromosome Location
- Chr. 14: 115044157 - 115044263 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029737.1 |
|
1 |
20 |
|
View Full Product Details