Gene Transcripts
- Gene Symbol
- Hspa13
- Entrez Gene ID
- 110920
- Gene Name
- heat shock protein 70 family, member 13
- Gene Aliases
- 1600002I10Rik, AV006182, B230217N24Rik, Stch
- Chromosome Location
- Chr. 16: 75755190 - 75767277 on Build GRCm38
- UniGene ID
- Mm.26704
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027492.1 |
|
5 |
834 |
|
GenBank mRNA |
AK030553.1 |
- |
5 |
807 |
|
BC094508.1 |
- |
5 |
777 |
|
View Full Product Details