Gene Transcripts
- Gene Symbol
- Mir7b
- Entrez Gene ID
- 723883
- Gene Name
- microRNA 7b
- Gene Aliases
- Mirn7b, mir-7b, mmu-mir-7b
- Chromosome Location
- Chr. 17: 56242988 - 56243098 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029827.1 |
|
1 |
54 |
|
View Full Product Details