Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NR_002870.2 | 1 | 4542 | |||
GenBank mRNA | AK051872.1 | - | LIT3445 D230012O04Rik |
1 | 550 | |
AK053654.1 | - | 1 | 3326 | |||
AK164873.1 | - | 1 | 2370 | |||
AY751519.1 | - | 1 | 4502 |