Gene Transcripts
- Gene Symbol
- Pradc1
- Entrez Gene ID
- 73327
- Gene Name
- protease-associated domain containing 1
- Gene Aliases
- 1700040I03Rik, Pap21
- Chromosome Location
- Chr. 6: 85446785 - 85451970 on Build GRCm38
- UniGene ID
- Mm.379221
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028081.1 |
|
1, 2 |
214 |
|
GenBank mRNA |
BG088874.2 |
- |
1, 2 |
189 |
|
CJ075544.1 |
- |
1, 2 |
214 |
|
View Full Product Details