Gene Transcripts
- Gene Symbol
- Bmp1
- Entrez Gene ID
- 12153
- Gene Name
- bone morphogenetic protein 1
- Gene Aliases
- Pcp, Tld
- Chromosome Location
- Chr. 14: 70474555 - 70520712 on Build GRCm38
- UniGene ID
- Mm.27757
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CX220176.1 |
- |
1 |
336 |
|
Chr. 14 - 70479602 - 70486621 [-] |
GenBankmRNA |
BQ917640.1 |
- |
1 |
371 |
|
Chr. 14 - 70479560 - 70486638 [-] |
View Full Product Details