Gene Transcripts
- Gene Symbol
- Tug1
- Entrez Gene ID
- 544752
- Gene Name
- taurine upregulated gene 1
- Gene Aliases
- AI316828
- Chromosome Location
- Chr. 11: 3639785 - 3648814 on Build GRCm38
- UniGene ID
- Mm.390314
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB245759.1 |
- |
1 |
75 |
|
Chr. 11 - 3645311 - 3647139 [-] |
GenBankmRNA |
CB244339.1 |
- |
1 |
75 |
|
Chr. 11 - 3645394 - 3647139 [-] |
GenBankmRNA |
CN535800.1 |
- |
1 |
28 |
|
Chr. 11 - 3645306 - 3647092 [-] |
View Full Product Details