Gene Transcripts

Gene Symbol
GUSBP11
Entrez Gene ID
91316
Gene Name
glucuronidase, beta pseudogene 11
Gene Aliases
Chromosome Location
Chr. 22: 23638487 - 23717423 on Build GRCh38
UniGene ID
Hs.148656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024448.2 8 1661
GenBank mRNA AK056875.1 - 8 1591
AK091837.1 - 7 1399
AK300413.1 - 7 1440
BX538181.1 - 7 1386

View Full Product Details