Gene Transcripts
- Gene Symbol
- IGLL3P
- Entrez Gene ID
- 91353
- Gene Name
- immunoglobulin lambda like polypeptide 3, pseudogene
- Gene Aliases
- 16.1, IGLL3
- Chromosome Location
- Chr. 22: 25318257 - 25320227 on Build GRCh38
- UniGene ID
- Hs.651077
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029395.1 |
|
1 |
71 |
|
- Gene Symbol
- GUSBP11
- Entrez Gene ID
- 91316
- Gene Name
- glucuronidase, beta pseudogene 11
- Gene Aliases
- Chromosome Location
- Chr. 22: 23638487 - 23717423 on Build GRCh38
- UniGene ID
- Hs.148656
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X52203.1 |
- |
1 |
407 |
|
View Full Product Details