Gene Transcripts
- Gene Symbol
- SNORA66
- Entrez Gene ID
- 26782
- Gene Name
- small nucleolar RNA, H/ACA box 66
- Gene Aliases
- RNU66, U66
- Chromosome Location
- Chr. 1: 92840719 - 92840851 on Build GRCh38
- UniGene ID
- Hs.180946
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002444.2 |
|
1 |
26 |
|
GenBank mRNA |
Y11160.1 |
- |
1 |
26 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
Z39353.1 |
- |
|
86 |
|
Chr. 5 - 19546118 - 19546133 [-] |
GenBankmRNA |
R49575.1 |
- |
|
95 |
|
Chr. - - [-] |
GenBankmRNA |
H17614.1 |
- |
1 |
91 |
|
Chr. 1 - 92840654 - 92841025 [+] |
View Full Product Details