Gene Transcripts
- Gene Symbol
- FAM95B1
- Entrez Gene ID
- 100133036
- Gene Name
- family with sequence similarity 95 member B1
- Gene Aliases
- Chromosome Location
- Chr. 9: 40323571 - 40329220 on Build GRCh38
- UniGene ID
- Hs.584289
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026759.1 |
|
1 |
139 |
|
GenBank mRNA |
AK125850.1 |
- |
1 |
2411 |
|
- Gene Symbol
- LOC105379252
- Entrez Gene ID
- 105379252
- Gene Name
- family with sequence similarity 95 member B1-like
- Gene Aliases
- Chromosome Location
- Chr. 9: 66047085 - 66052734 on Build GRCh38
- UniGene ID
- Hs.584289
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136310.1 |
|
1 |
139 |
|
View Full Product Details