Gene Transcripts
- Gene Symbol
- LRRC75A-AS1
- Entrez Gene ID
- 125144
- Gene Name
- LRRC75A antisense RNA 1
- Gene Aliases
- C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
- Chromosome Location
- Chr. 17: 16438987 - 16470648 on Build GRCh38
- UniGene ID
- Hs.368934
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AL556069.3 |
- |
1 |
14 |
|
Chr. 17 - 16438987 - 16442018 [+] |
GenBankmRNA |
BU600162.1 |
- |
1 |
18 |
|
Chr. 17 - 16438983 - 16441742 [+] |
GenBankmRNA |
BP230392.1 |
- |
1 |
15 |
|
Chr. 17 - 16438986 - 16441719 [+] |
View Full Product Details