Gene Transcripts

Gene Symbol
TCL6
Entrez Gene ID
27004
Gene Name
T-cell leukemia/lymphoma 6 (non-protein coding)
Gene Aliases
TNG1, TNG2
Chromosome Location
Chr. 14: 95651178 - 95673452 on Build GRCh38
UniGene ID
Hs.510368
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028288.1 1 1025
GenBank mRNA AB035337.1 - 1 1027
AB035338.1 - 1 1027
BX161462.1 - 2 690

View Full Product Details