Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_026752.1 | 7 | 1266 | ||
GenBank mRNA | AK128484.1 | - | 1577 | ||
BC006312.2 | - | 7 | 1520 | ||
BC010508.1 | - | 4 | 791 | ||
BC047736.1 | - | 7 | 1266 | ||
DQ786196.1 | - | 2 | 679 |