Gene Transcripts
- Gene Symbol
- AATK-AS1
- Entrez Gene ID
- 388428
- Gene Name
- AATK antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 17: 81165507 - 81183164 on Build GRCh38
- UniGene ID
- Hs.591202
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027255.1 |
|
6 |
1543 |
|
GenBank mRNA |
AK126811.1 |
- |
6 |
1543 |
|
View Full Product Details