Gene Transcripts

Gene Symbol
EGFEM1P
Entrez Gene ID
93556
Gene Name
EGF like and EMI domain containing 1, pseudogene
Gene Aliases
C3orf50, NCRNA00259
Chromosome Location
Chr. 3: 168249522 - 168830599 on Build GRCh38
UniGene ID
Hs.478158
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_021485.2 16 2682
GenBank mRNA BC011266.2 - 6 978
BC103698.1 - 4 799
BC104748.1 - 4 798
BC105690.1 - 4 798

View Full Product Details