Gene Transcripts
- Gene Symbol
- COPG2IT1
- Entrez Gene ID
- 53844
- Gene Name
- COPG2 imprinted transcript 1 (non-protein coding)
- Gene Aliases
- CIT1, COPG2AS, DKFZP761N09121, NCRNA00170
- Chromosome Location
- Chr. 7: 130543453 - 130546900 on Build GRCh38
- UniGene ID
- Hs.6421
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024086.2 |
|
1 |
2805 |
|
GenBank mRNA |
AF038190.1 |
- |
1 |
1247 |
|
AK123640.1 |
- |
1 |
2800 |
|
View Full Product Details