Gene Transcripts
- Gene Symbol
- HAS2-AS1
- Entrez Gene ID
- 594842
- Gene Name
- HAS2 antisense RNA 1
- Gene Aliases
- HAS2-AS, HAS2AS, HASNT, NCRNA00077
- Chromosome Location
- Chr. 8: 121639346 - 121645325 on Build GRCh38
- UniGene ID
- Hs.668686
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002835.2 |
|
4 |
1462 |
|
GenBank mRNA |
AY941178.1 |
- |
4 |
1462 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CA336013.1 |
- |
3 |
401 |
|
Chr. 8 - 121641436 - 121644693 [+] |
View Full Product Details