Gene Transcripts
- Gene Symbol
- DSCR10
- Entrez Gene ID
- 259234
- Gene Name
- Down syndrome critical region 10 (non-protein coding)
- Gene Aliases
- Chromosome Location
- Chr. 21: 38206156 - 38208644 on Build GRCh38
- UniGene ID
- Hs.147695
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027695.1 |
|
1 |
61 |
|
GenBank mRNA |
AB066291.1 |
- |
1 |
61 |
|
BX115939.1 |
- |
1 |
29 |
|
View Full Product Details