Gene Transcripts
- Gene Symbol
- MIR34A
- Entrez Gene ID
- 407040
- Gene Name
- microRNA 34a
- Gene Aliases
- MIRN34A, miRNA34A, mir-34, mir-34a
- Chromosome Location
- Chr. 1: 9151668 - 9151777 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029610.1 |
|
1 |
68 |
|
- Gene Symbol
- MIR34AHG
- Entrez Gene ID
- 106614088
- Gene Name
- MIR34A host gene
- Gene Aliases
- Chromosome Location
- Chr. 1: 9148011 - 9182338 on Build GRCh38
- UniGene ID
- Hs.551837
- Species
- Homo sapiens
View Full Product Details