Gene Transcripts

Gene Symbol
TMEM41B
Entrez Gene ID
440026
Gene Name
transmembrane protein 41B
Gene Aliases
Chromosome Location
Chr. 11: 9280654 - 9314768 on Build GRCh38
UniGene ID
Hs.594563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015012.3 NP_055827.1 7 3864
NR_028491.2 8 1535
GenBank mRNA BC035034.1 - 7 3742
BC044597.1 - 7 3732 4811565
BC091524.1 - 8 1334 30512350

View Full Product Details