Gene Transcripts
- Gene Symbol
- PRINS
- Entrez Gene ID
- 100169750
- Gene Name
- psoriasis associated non-protein coding RNA induced by stress
- Gene Aliases
- NCRNA00074
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.731140
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022045.1 |
- |
LIT3418 PRINS |
2 |
1956 |
|
View Full Product Details