Gene Transcripts
- Gene Symbol
- CTSL3P
- Entrez Gene ID
- 392360
- Gene Name
- cathepsin L family member 3, pseudogene
- Gene Aliases
- CTSL3, HCTSL-s
- Chromosome Location
- Chr. 9: 87772915 - 87786884 on Build GRCh38
- UniGene ID
- Hs.690450
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027917.1 |
|
3 |
256 |
|
GenBank mRNA |
AJ851862.1 |
- |
3 |
256 |
|
View Full Product Details