Gene Transcripts
- Gene Symbol
- TCAM1P
- Entrez Gene ID
- 146771
- Gene Name
- testicular cell adhesion molecule 1, pseudogene
- Gene Aliases
- TCAM1
- Chromosome Location
- Chr. 17: 63857016 - 63864379 on Build GRCh38
- UniGene ID
- Hs.629177
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002947.2 |
|
1 |
158 |
|
GenBank mRNA |
AB026156.1 |
- |
1 |
158 |
|
LC005096.1 |
- |
3 |
249 |
|
View Full Product Details