Gene Transcripts
- Gene Symbol
- RNF217-AS1
- Entrez Gene ID
- 7955
- Gene Name
- RNF217 antisense RNA 1 (head to head)
- Gene Aliases
- STL
- Chromosome Location
- Chr. 6: 124908243 - 124963027 on Build GRCh38
- UniGene ID
- Hs.544438
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026876.1 |
|
7 |
1616 |
|
GenBank mRNA |
AL713649.2 |
- |
7 |
1616 |
|
View Full Product Details