Gene Transcripts

Gene Symbol
TUG1
Entrez Gene ID
55000
Gene Name
taurine up-regulated 1 (non-protein coding)
Gene Aliases
LINC00080, NCRNA00080, TI-227H
Chromosome Location
Chr. 22: 30969211 - 30979395 on Build GRCh38
UniGene ID
Hs.554829
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002323.2 3 5278
NR_110492.1 4 5334
NR_110493.1 3 5264
GenBank mRNA AB073661.1 - 1 1733
CB127883.1 - 1 95

View Full Product Details