Gene Transcripts
- Gene Symbol
- SNRPD3
- Entrez Gene ID
- 6634
- Gene Name
- small nuclear ribonucleoprotein D3 polypeptide
- Gene Aliases
- SMD3, Sm-D3
- Chromosome Location
- Chr. 22: 24555650 - 24574964 on Build GRCh38
- UniGene ID
- Hs.356549
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR591143.1 |
- |
1 |
168 |
|
Chr. 22 - 24573938 - 24574894 [+] |
GenBankmRNA |
AV718073.1 |
- |
1 |
188 |
|
Chr. 22 - 24573918 - 24574599 [+] |
GenBankmRNA |
BE735004.1 |
- |
1 |
241 |
|
Chr. 22 - 24573905 - 24574771 [+] |
GenBankmRNA |
BX432338.2 |
- |
1 |
168 |
|
Chr. 22 - 24573938 - 24574894 [+] |
GenBankmRNA |
BX432339.2 |
- |
1 |
168 |
|
Chr. 22 - 24573938 - 24574752 [+] |
View Full Product Details