Gene Transcripts
- Gene Symbol
- CXorf23
- Entrez Gene ID
- 256643
- Gene Name
- chromosome X open reading frame 23
- Gene Aliases
- Chromosome Location
- Chr. X: 19912860 - 19991040 on Build GRCh38
- UniGene ID
- Hs.28896
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC093092.1 |
- |
1 |
595 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CD102745.1 |
- |
1 |
597 |
|
Chr. X - 19965743 - 19966464 [-] |
View Full Product Details