Gene Transcripts
- Gene Symbol
- GALNT18
- Entrez Gene ID
- 374378
- Gene Name
- polypeptide N-acetylgalactosaminyltransferase 18
- Gene Aliases
- GALNACT18, GALNT15, GALNTL4, GalNAc-T15, GalNAc-T18
- Chromosome Location
- Chr. 11: 11270874 - 11622045 on Build GRCh38
- UniGene ID
- Hs.655152
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124690.1 |
- |
1 |
1279 |
|
View Full Product Details