Gene Transcripts
- Gene Symbol
- CDX2
- Entrez Gene ID
- 1045
- Gene Name
- caudal type homeobox 2
- Gene Aliases
- CDX-3, CDX2/AS, CDX3
- Chromosome Location
- Chr. 13: 27960918 - 27969368 on Build GRCh38
- UniGene ID
- Hs.127383
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK000205.1 |
- |
1 |
188 |
|
View Full Product Details