Gene Transcripts
- Gene Symbol
- TMEM59
- Entrez Gene ID
- 9528
- Gene Name
- transmembrane protein 59
- Gene Aliases
- C1orf8, DCF1, HSPC001, PRO195, UNQ169
- Chromosome Location
- Chr. 1: 54026681 - 54053573 on Build GRCh38
- UniGene ID
- Hs.118947
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK129688.1 |
- |
1 |
714 |
|
View Full Product Details