Gene Transcripts
- Gene Symbol
- TFDP2
- Entrez Gene ID
- 7029
- Gene Name
- transcription factor Dp-2
- Gene Aliases
- DP2
- Chromosome Location
- Chr. 3: 141944428 - 142149544 on Build GRCh38
- UniGene ID
- Hs.379018
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB133997.1 |
- |
1 |
497 |
|
Chr. 3 - 141947603 - 141948143 [-] |
GenBankmRNA |
CN417698.1 |
- |
1 |
231 |
|
Chr. 3 - 141947434 - 141947877 [-] |
GenBankmRNA |
HY056991.1 |
- |
1 |
352 |
|
Chr. 3 - 141947522 - 141947998 [-] |
View Full Product Details