Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005257083.2 | XP_005257140.1 | 8 | 7245 | |
XM_005257084.2 | XP_005257141.1 | 8 | 7203 | ||
XM_011524412.1 | XP_011522714.1 | 7 | 7144 | ||
XM_011524413.1 | XP_011522715.1 | 7 | 7125 | ||
GenBank mRNA | BC030561.1 | - | 1 | 1238 |