Gene Transcripts
- Gene Symbol
- GGACT
- Entrez Gene ID
- 87769
- Gene Name
- gamma-glutamylamine cyclotransferase
- Gene Aliases
- A2LD1
- Chromosome Location
- Chr. 13: 100530164 - 100588792 on Build GRCh38
- UniGene ID
- Hs.350868
- Species
- Homo sapiens
- Gene Symbol
- LOC100507654
- Entrez Gene ID
- 100507654
- Gene Name
- uncharacterized LOC100507654
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.651545
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC034289.1 |
- |
4 |
1830 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000454752.3 |
- |
4 |
1826 |
|
Chr. 13 - 100535741 - 100580117 [-] |
View Full Product Details