Gene Transcripts
- Gene Symbol
- EEF1D
- Entrez Gene ID
- 1936
- Gene Name
- eukaryotic translation elongation factor 1 delta
- Gene Aliases
- EF-1D, EF1D, FP1047
- Chromosome Location
- Chr. 8: 143579697 - 143597675 on Build GRCh38
- UniGene ID
- Hs.333388
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC046445.1 |
- |
1 |
2555 |
|
View Full Product Details