Gene Transcripts
- Gene Symbol
- DYSF
- Entrez Gene ID
- 8291
- Gene Name
- dysferlin
- Gene Aliases
- FER1L1, LGMD2B, MMD1
- Chromosome Location
- Chr. 2: 71453155 - 71686763 on Build GRCh38
- UniGene ID
- Hs.252180
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK074442.1 |
- |
1 |
936 |
|
View Full Product Details